accessibility menu, dialog, popup

UserName

Viewing profile as user:
H2CO

CAS RN 85721-25-1

IUPAC Name: 2-(oct-7-en-1-yl)oxirane
Synonyms: 2-(oct-7-en-1-yl)oxirane
Molecular Weight (g/mol): 154.25
Molecular Formula: C10H18O
InChi Key: FCZHJHKCOZGQJZ-UHFFFAOYNA-N
SMILES: C=CCCCCCCC1CO1
Molecular Weight (g/mol) 
  • (1)
  • (1)
Percent Purity 
  • (1)
  • (1)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)