CAS RN 85722-06-1
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
1,1'-Dipropylhafnocene Dichloride 98.0+%, TCI America™
CAS: 85722-06-1 Molecular Formula: C16H22Cl2Hf Synonym: Bis(propylcyclopentadienyl)hafnium(IV) Dichloride