Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1,1'-Dipropylhafnocene Dichloride 98.0+%, TCI America™
SDP

Catalog No. D32861G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D32861G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. D32861G Supplier TCI America Supplier No. D32861G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 85722-06-1
Molecular Formula C16H22Cl2Hf
Synonym Bis(propylcyclopentadienyl)hafnium(IV) Dichloride

Specifications

Quantity 1 g
Formula Weight 463.74
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material 1,1′-Dipropylhafnocene Dichloride
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.