accessibility menu, dialog, popup

UserName

Viewing profile as user:
ONOOH

CAS RN 857283-84-2

IUPAC Name: 5-(furan-2-yl)pyridine-3-carboxylic acid
Synonyms: 5-(furan-2-yl)pyridine-3-carboxylic acid
Molecular Weight (g/mol): 189.17
Molecular Formula: C10H7NO3
InChi Key: YAIWRLRMUNJGCX-UHFFFAOYSA-N
SMILES: OC(=O)C1=CN=CC(=C1)C1=CC=CO1
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results