CAS RN 857478-30-9
CAS RN 857478-30-9
IUPAC Name:
2-({[(9H-fluoren-9-yl)methoxy]carbonyl}amino)-2-methylbutanoic acid
Synonyms:
2-{[(9H-fluoren-9-ylmethoxy)carbonyl]amino}-2-methylbutanoic acid
Molecular Weight (g/mol):
339.39
Molecular Formula:
C20H21NO4
InChi Key:
DZSLHAJXIQCMLR-FQEVSTJZSA-N
SMILES:
CCC(C)(NC(=O)OCC1C2=C(C=CC=C2)C2=C1C=CC=C2)C(O)=O
1
–
1
of
1
results
1
Advanced Chem Tech Fmoc-Iva-OH, Advanced ChemTech
CAS: 857478-30-9 Molecular Formula: C20H21NO4 Molecular Weight (g/mol): 339.39 MDL Number: MFCD12031688 InChI Key: DZSLHAJXIQCMLR-FQEVSTJZSA-N IUPAC Name: 2-({[(9H-fluoren-9-yl)methoxy]carbonyl}amino)-2-methylbutanoic acid SMILES: CCC(C)(NC(=O)OCC1C2=C(C=CC=C2)C2=C1C=CC=C2)C(O)=O