accessibility menu, dialog, popup

UserName

Viewing profile as user:
OHNH2

CAS RN 857831-26-6

IUPAC Name: 2-(4-aminocyclohexyl)ethan-1-ol
Synonyms: 2-(4-aminocyclohexyl)ethanol
Molecular Weight (g/mol): 143.23
Molecular Formula: C8H17NO
InChi Key: XLKSLYGRARRUJP-UHFFFAOYSA-N
SMILES: NC1CCC(CCO)CC1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results