Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-Aminocyclohexaneethanol (cis- and trans- mixture) 98.0+%, TCI America™
SDP

Catalog No. A22885G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A22885G 5 g
A22881G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. A22885G Supplier TCI America Supplier No. A22885G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 857831-26-6
Molecular Formula C8H17NO
Molecular Weight (g/mol) 143.23
InChI Key XLKSLYGRARRUJP-UHFFFAOYSA-N
Synonym 2-(4-Aminocyclohexyl)ethanol, 1-Amino-4-(2-hydroxyethyl)cyclohexane
PubChem CID 21312787
IUPAC Name 2-(4-aminocyclohexyl)ethanol
SMILES C1CC(CCC1CCO)N

Specifications

Melting Point 85°C
Color White
Quantity 5 g
Formula Weight 143.23
Percent Purity ≥98.0% (T)
Physical Form Crystals
Chemical Name or Material 4-Aminocyclohexaneethanol (cis- and trans- mixture)
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.