accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3ONH3COOCH3CH3CH3CH3OCl(E)(E)

CAS RN 85801-02-1

IUPAC Name: (2E,6E)-3,7,11-trimethyldodeca-2,6,10-trien-1-yl 2-[1-(4-chlorobenzoyl)-5-methoxy-2-methyl-1H-indol-3-yl]acetate
Synonyms: infree indomethacin farnesil IMF C34H40ClNO4 indometacin farnesil
Molecular Weight (g/mol): 562.15
Molecular Formula: C34H40ClNO4
InChi Key: CFIGYZZVJNJVDQ-LMJOQDENSA-N
SMILES: COC1=CC=C2N(C(=O)C3=CC=C(Cl)C=C3)C(C)=C(CC(=O)OC\C=C(/C)CC\C=C(/C)CCC=C(C)C)C2=C1
Percent Purity 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results