Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Indometacin Farnesil 98.0+%, TCI America™
SDP

Catalog No. I0943100MG Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 mg
100 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
I0943100MG 100 mg
I094325MG 25 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. I0943100MG Supplier TCI America Supplier No. I0943100MG
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 85801-02-1
Molecular Formula C34H40ClNO4
Molecular Weight (g/mol) 562.147
MDL Number MFCD00870661
InChI Key CFIGYZZVJNJVDQ-LMJOQDENSA-N
PubChem CID 5282183
IUPAC Name [(2E,6E)-3,7,11-trimethyldodeca-2,6,10-trienyl] 2-[1-(4-chlorobenzoyl)-5-methoxy-2-methylindol-3-yl]acetate
SMILES CC1=C(C2=C(N1C(=O)C3=CC=C(C=C3)Cl)C=CC(=C2)OC)CC(=O)OCC=C(C)CCC=C(C)CCC=C(C)C

Specifications

Color Yellow
Quantity 100 mg
Formula Weight 562.15
Percent Purity ≥98.0% (HPLC)
Physical Form Liquid
Chemical Name or Material Indometacin Farnesil
RTECSNumber NL3522820
TSCA No
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.