accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 85916-13-8

H3CCH3H3COOHNOOCH3CH3H3C

CAS RN 85916-13-8

IUPAC Name: tert-butyl 2-{[2-(tert-butoxy)-2-oxoethyl]amino}acetate
Synonyms: tert-butyl 2-{[2-(tert-butoxy)-2-oxoethyl]amino}acetate
Molecular Weight (g/mol): 245.32
Molecular Formula: C12H23NO4
InChi Key: SMXMBXPLRFTROI-UHFFFAOYSA-N
SMILES: CC(C)(C)OC(=O)CNCC(=O)OC(C)(C)C
Percent Purity 
  • (2)
  • (1)
Quantity 
  • (2)
  • (1)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)
  • (2)
  • (2)
  • (2)
  • (1)
  • (2)
  • (1)
  • (2)