Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Di-tert-butyl Iminodiacetate 97.0+%, TCI America™
SDP

Catalog No. D41095G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D41095G 5 g
D41091G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D41095G Supplier TCI America Supplier No. D41095G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 85916-13-8
Molecular Formula C12H23NO4
Molecular Weight (g/mol) 245.32
MDL Number MFCD01863672
InChI Key SMXMBXPLRFTROI-UHFFFAOYSA-N
Synonym Iminodiacetic Acid Di-tert-butyl Ester
PubChem CID 4393691
IUPAC Name tert-butyl 2-{[2-(tert-butoxy)-2-oxoethyl]amino}acetate
SMILES CC(C)(C)OC(=O)CNCC(=O)OC(C)(C)C

Specifications

Melting Point 42°C
Color White
Quantity 5 g
Formula Weight 245.32
Percent Purity ≥97.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material Di-tert-butyl Iminodiacetate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.