accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 859212-16-1

H3CNHONNCH3H3CFFFNHNNNN

CAS RN 859212-16-1

IUPAC Name: N-[3-({[4,5'-bipyrimidin]-2-yl}amino)-4-methylphenyl]-4-{[3-(dimethylamino)pyrrolidin-1-yl]methyl}-3-(trifluoromethyl)benzamide
Synonyms: N-(3-{[4,5'-bipyrimidin]-2-ylamino}-4-methylphenyl)-4-{[3-(dimethylamino)pyrrolidin-1-yl]methyl}-3-(trifluoromethyl)benzamide
Molecular Weight (g/mol): 576.63
Molecular Formula: C30H31F3N8O
InChi Key: ZGBAJMQHJDFTQJ-UHFFFAOYNA-N
SMILES: CN(C)C1CCN(CC2=C(C=C(C=C2)C(=O)NC2=CC(NC3=NC=CC(=N3)C3=CN=CN=C3)=C(C)C=C2)C(F)(F)F)C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results