accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3OOFCl

CAS RN 85953-29-3

IUPAC Name: methyl 2-chloro-4-fluorobenzoate
Synonyms: methyl 2-chloro-4-fluorobenzoate
Molecular Weight (g/mol): 188.58
Molecular Formula: C8H6ClFO2
InChi Key: YZDLPZNWBRBZMZ-UHFFFAOYSA-N
SMILES: COC(=O)C1=CC=C(F)C=C1Cl
Percent Purity 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results