Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Methyl 2-Chloro-4-fluorobenzoate 98.0+%, TCI America™
SDP

Catalog No. M31025G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M31025G 5 g
M310225G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. M31025G Supplier TCI America Supplier No. M31025G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 85953-29-3
Molecular Formula C8H6ClFO2
Molecular Weight (g/mol) 188.582
MDL Number MFCD00173937
InChI Key YZDLPZNWBRBZMZ-UHFFFAOYSA-N
Synonym 2-chloro-4-fluorobenzoic acid methyl ester, methyl 2-chloro-4-fluorobenzenecarboxylate, benzoic acid, 2-chloro-4-fluoro-, methyl ester, 2-chloro-4-fluoro-benzoic acid methyl ester, pubchem18760, rarechem al bf 0289, acmc-209q7x, ksc495c1p, methyl 2-chloro-4-fluorob..., methy 2-chloro-4-fluorobenzoate
PubChem CID 2763551
IUPAC Name methyl 2-chloro-4-fluorobenzoate
SMILES COC(=O)C1=C(C=C(C=C1)F)Cl

Specifications

Color Yellow
Boiling Point 105°C
Quantity 5 g
Formula Weight 188.58
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material Methyl 2-Chloro-4-fluorobenzoate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.