accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 85958-57-2

IUPAC Name: 2-chloro-4-fluorobenzene-1-sulfonyl chloride
Synonyms: 2-chloro-4-fluorobenzenesulfonyl chloride
Molecular Weight (g/mol): 229.05
Molecular Formula: C6H3Cl2FO2S
InChi Key: FCFPJKHVCHKCMP-UHFFFAOYSA-N
SMILES: FC1=CC(Cl)=C(C=C1)S(Cl)(=O)=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results