CAS RN 859850-90-1
IUPAC Name:
3-[(4-methylpiperazin-1-yl)methyl]benzonitrile
Synonyms:
3-[(4-methylpiperazin-1-yl)methyl]benzonitrile
Molecular Weight (g/mol):
215.3
Molecular Formula:
C13H17N3
InChi Key:
NNYDAONDPGWBRI-UHFFFAOYSA-N
SMILES:
CN1CCN(CC2=CC=CC(=C2)C#N)CC1
1
–
1
of
1
results
1
3-[(4-Methylpiperazin-1-yl)methyl]benzonitrile, ≥97%, Thermo Scientific™
CAS: 859850-90-1 Molecular Formula: C13H17N3 Molecular Weight (g/mol): 215.30 MDL Number: MFCD07772861 InChI Key: NNYDAONDPGWBRI-UHFFFAOYSA-N Synonym: 3-4-methylpiperazin-1-yl methyl benzonitrile,3-4-methyl-piperazin-1-yl-methyl-benzonitrile,3-4-methyl-piperazin-1-ylmethyl-benzonitrile,3-4-methylpiperazinyl methyl benzenecarbonitrile,benzonitrile,3-4-methyl-1-piperazinyl methyl PubChem CID: 7164647 IUPAC Name: 3-[(4-methylpiperazin-1-yl)methyl]benzonitrile SMILES: CN1CCN(CC2=CC=CC(=C2)C#N)CC1