accessibility menu, dialog, popup

UserName

Viewing profile as user:
NNNO

CAS RN 859850-94-5

IUPAC Name: 4-[(1H-1,2,4-triazol-1-yl)methyl]benzaldehyde
Synonyms: 4-(1,2,4-triazol-1-ylmethyl)benzaldehyde
Molecular Weight (g/mol): 187.2
Molecular Formula: C10H9N3O
InChi Key: XEPIORKIFKJATD-UHFFFAOYSA-N
SMILES: O=CC1=CC=C(CN2C=NC=N2)C=C1
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results