Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-(1H-1,2,4-Triazol-1-ylmethyl)benzaldehyde 97+%, Thermo Scientific™

Catalog No. CC45704DE
Change view
Click to view available options
Quantity:
250 mg
1 g
5 g
Packaging:
Amber Glass Bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
CC45704DE 5 g Amber Glass Bottle
CC45704CB 250 mg Amber Glass Bottle
CC45704DA 1 g Amber Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. CC45704DE Supplier Thermo Scientific Chemicals Supplier No. CC45704DE
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 859850-94-5
Molecular Formula C10H9N3O
Molecular Weight (g/mol) 187.202
InChI Key XEPIORKIFKJATD-UHFFFAOYSA-N
Synonym 4-1h-1,2,4-triazol-1-ylmethyl benzaldehyde, 4-1,2,4-triazol-1-ylmethyl benzaldehyde, 4-1,2,4-triazolylmethyl benzaldehyde, 4-1h-1,2,4-triazol-1-yl methyl benzaldehyde, benzaldehyde,4-1h-1,2,4-triazol-1-ylmethyl
PubChem CID 7164655
IUPAC Name 4-(1,2,4-triazol-1-ylmethyl)benzaldehyde
SMILES C1=CC(=CC=C1CN2C=NC=N2)C=O

Specifications

Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 5 g
Formula Weight 187.2
Percent Purity 97%
Chemical Name or Material 4-(1H-1, 2, 4-Triazol-1-ylmethyl)benzaldehyde
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.