accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 86-26-0

IUPAC Name: 2-methoxy-1,1'-biphenyl
Synonyms: 1,1'-biphenyl, 2-methoxy-
Molecular Weight (g/mol): 184.24
Molecular Formula: C13H12O
InChi Key: NLWCWEGVNJVLAX-UHFFFAOYSA-N
SMILES: COC1=CC=CC=C1C1=CC=CC=C1
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results