Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Methoxybiphenyl 98.0+%, TCI America™
SDP

Catalog No. M171325G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M171325G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. M171325G Supplier TCI America Supplier No. M171325G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 86-26-0
Molecular Formula C13H12O
Molecular Weight (g/mol) 184.24
MDL Number MFCD00008367
InChI Key NLWCWEGVNJVLAX-UHFFFAOYSA-N
Synonym 2-methoxybiphenyl, 2-phenylanisole, 1,1'-biphenyl, 2-methoxy, o-phenylanisole, anisole, o-phenyl, methyl diphenyl ether, o-phenyl anisole, biphenyl-2-yl methyl ether, 2-methoxy-1,1'-biphenyl, 2-methoxy-biphenylate
PubChem CID 6835
IUPAC Name 2-methoxy-1,1'-biphenyl
SMILES COC1=CC=CC=C1C1=CC=CC=C1

Specifications

Melting Point 28°C
Color White-Yellow
Boiling Point 274°C
Quantity 25 g
Formula Weight 184.24
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Lumps
Chemical Name or Material 2-Methoxybiphenyl
EINECSNumber (4)-0056
RTECSNumber BZ8835000
TSCA Yes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.