accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 86-50-0

IUPAC Name: O,O-dimethyl {[(4-oxo-3,4-dihydro-1,2,3-benzotriazin-3-yl)methyl]sulfanyl}phosphonothioate
Synonyms: azinphos azinfos-methyl metazintox azinphos methyl caswell no. 374 guthion gusathion methyl epa shaughnessy #058001 benzotriazinedithiophosphoric acid dimethoxy ester cotnion
Molecular Weight (g/mol): 317.32
Molecular Formula: C10H12N3O3PS2
InChi Key: CJJOSEISRRTUQB-UHFFFAOYSA-N
SMILES: COP(=S)(OC)SCN1N=NC2=CC=CC=C2C1=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results