Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Guthion™ Azinphos-methyl, MilliporeSigma™ Supelco™

Catalog No. 111020208 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
250 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
11-102-0208 250 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 11-102-0208 Supplier MilliporeSigma Supelco Supplier No. 45333250MG
Add to Cart
Add to Cart
Agriculture and Environmental
TRUSTED_SUSTAINABILITY

Specifications

Product Line Guthion
CAS 86-50-0
Physical Form Neat
Linear Formula C10H12N3O3 PS2
MDL Number MFCD00041814
Synonym Guthion(R); Methyltriazotin
Type Analytical Standard
Quantity 250 mg
Formula Weight 317.32
Grade Analytical Standard

Guthion is a registered trademark of Makhteshim Chemical Works Ltd.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.