CAS RN 86060-92-6
CAS RN 86060-92-6
IUPAC Name:
2,3,4,5,6-pentafluorophenyl 2-({[(9H-fluoren-9-yl)methoxy]carbonyl}amino)-3-phenylpropanoate
Synonyms:
2,3,4,5,6-pentafluorophenyl 2-{[(9H-fluoren-9-ylmethoxy)carbonyl]amino}-3-phenylpropanoate
Molecular Weight (g/mol):
553.49
Molecular Formula:
C30H20F5NO4
InChi Key:
WKHPSOMXNCTXPK-UHFFFAOYNA-N
SMILES:
FC1=C(F)C(F)=C(OC(=O)C(CC2=CC=CC=C2)NC(=O)OCC2C3=CC=CC=C3C3=CC=CC=C23)C(F)=C1F
1
–
1
of
1
results
1
Advanced Chem Tech Fmoc-Phe-OPfp, Advanced ChemTech
CAS: 86060-92-6 Molecular Formula: C30H20F5NO4 Molecular Weight (g/mol): 553.49 InChI Key: WKHPSOMXNCTXPK-UHFFFAOYNA-N IUPAC Name: 2,3,4,5,6-pentafluorophenyl 2-({[(9H-fluoren-9-yl)methoxy]carbonyl}amino)-3-phenylpropanoate SMILES: FC1=C(F)C(F)=C(OC(=O)C(CC2=CC=CC=C2)NC(=O)OCC2C3=CC=CC=C3C3=CC=CC=C23)C(F)=C1F