accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3CCH3NH2

CAS RN 86089-17-0

IUPAC Name: 8-ethylundecan-1-amine
Synonyms: 8-ethylundecan-1-amine
Molecular Weight (g/mol): 199.38
Molecular Formula: C13H29N
InChi Key: KFBNTNYCQVDLBK-UHFFFAOYNA-N
SMILES: CCCC(CC)CCCCCCCN
Percent Purity 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results