Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tridecylamine (=Monotridecylamine) (mixture of branched chain isomers) 98.0+%, TCI America™
SDP

Catalog No. T0936500ML Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 mL
500 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T0936500ML 500 mL
T093625ML 25 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. T0936500ML Supplier TCI America Supplier No. T0936500ML
Add to Cart
Add to Cart

Chemical Identifiers

CAS 86089-17-0
Molecular Formula C13H29N
MDL Number MFCD00008156

Specifications

Color Yellow
Quantity 500 mL
UN Number 2735
Formula Weight 199.38
Percent Purity ≥98.0% (T)
Physical Form Liquid
Chemical Name or Material Tridecylamine (=Monotridecylamine) (mixture of branched chain isomers)
EINECSNumber (2)-0133
RTECSNumber YD3011650
TSCA Yes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.