accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH2NH2HCl

CAS RN 86128-83-8

IUPAC Name: tricyclo[3.3.1.0³,⁷]nonan-3-amine hydrochloride
Synonyms: tricyclo[3.3.1.0³,⁷]nonan-3-amine hydrochloride
Molecular Weight (g/mol): 173.68
Molecular Formula: C9H16ClN
InChi Key: YPEGOYFGBDTDJK-UHFFFAOYNA-N
SMILES: Cl.NC12CC3CC1CC(C2)C3
Percent Purity 
  • (2)
  • (1)
Physical Form 
  • (2)
  • (1)
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (2)
  • (1)
  • (2)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)