Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3-Noradamantanamine Hydrochloride 98.0+%, TCI America™
SDP

Catalog No. N07785G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
N07785G 5 g
N07781G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. N07785G Supplier TCI America Supplier No. N07785G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 86128-83-8
Molecular Formula C9H16ClN
Molecular Weight (g/mol) 173.684
MDL Number MFCD00213499
InChI Key YPEGOYFGBDTDJK-UHFFFAOYSA-N
Synonym 3-noradamantanamine hydrochloride, octahydro-2,5-methanopentalen-3a-amine hydrochloride, 3-aminonoradamantane hcl, 3-aminonoradamantane hydrochloride, tricyclo 3.3.1.0 3 ,? nonan-3-amine hydrochloride, 3-noradamantanamine hcl, c9h15n.hcl, 3-noradamantanaminehydrochloride, 3-noradamantanamine hydrochloride 95
PubChem CID 12791864
SMILES C1C2CC3CC1CC3(C2)N.Cl

Specifications

Melting Point 350°C
Color White-Yellow
Quantity 5 g
Formula Weight 173.68
Percent Purity ≥98.0% (N,T)
Physical Form Crystalline Powder
Chemical Name or Material 3-Noradamantanamine Hydrochloride
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.