accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3COOClCH3NCl

CAS RN 86129-63-7

IUPAC Name: ethyl 2,4-dichloro-6-methylpyridine-3-carboxylate
Synonyms: ethyl 2,4-dichloro-6-methylpyridine-3-carboxylate
Molecular Weight (g/mol): 234.08
Molecular Formula: C9H9Cl2NO2
InChi Key: ZNFJVVLTQSOWJY-UHFFFAOYSA-N
SMILES: CCOC(=O)C1=C(Cl)C=C(C)N=C1Cl

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results