accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3CCH2OSiSiCH3CH3CH3SiCH3CH3H3CSiCH3CH3H3C

CAS RN 861869-12-7

IUPAC Name: 1,1,1,3,3,3-hexamethyl-2-(prop-1-en-2-yloxy)-2-(trimethylsilyl)trisilane
Synonyms: 1,1,1,3,3,3-hexamethyl-2-(prop-1-en-2-yloxy)-2-(trimethylsilyl)trisilane
Molecular Weight (g/mol): 304.73
Molecular Formula: C12H32OSi4
InChi Key: PAQMOJBNTFYAFL-UHFFFAOYSA-N
SMILES: CC(=C)O[Si]([Si](C)(C)C)([Si](C)(C)C)[Si](C)(C)C
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)
  • (1)
  • (1)
  • (1)