Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Isopropenyloxytris(trimethylsilyl)silane 94.0+%, TCI America™
SDP

Catalog No. I08851G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
I08851G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. I08851G Supplier TCI America Supplier No. I08851G

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 861869-12-7
Molecular Formula C12H32OSi4
Molecular Weight (g/mol) 304.727
InChI Key PAQMOJBNTFYAFL-UHFFFAOYSA-N
PubChem CID 54762912
IUPAC Name trimethyl-[prop-1-en-2-yloxy-bis(trimethylsilyl)silyl]silane
SMILES CC(=C)O[Si]([Si](C)(C)C)([Si](C)(C)C)[Si](C)(C)C

Specifications

Color White
Quantity 1 g
Formula Weight 304.73
Percent Purity ≥94.0% (GC)
Physical Form Crystalline Lumps
Chemical Name or Material Isopropenyloxytris(trimethylsilyl)silane
TSCA No
Recommended Storage Refrigerator
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.