accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 861891-50-1

IUPAC Name: 1-methyl-3-(1-methyl-1H-indol-3-yl)-4-(pentylamino)-2,5-dihydro-1H-pyrrole-2,5-dione
Synonyms: 1-methyl-3-(1-methylindol-3-yl)-4-(pentylamino)pyrrole-2,5-dione
Molecular Weight (g/mol): 325.41
Molecular Formula: C19H23N3O2
InChi Key: SGLOMINNEBLJFF-UHFFFAOYSA-N
SMILES: CCCCCNC1=C(C(=O)N(C)C1=O)C1=CN(C)C2=CC=CC=C12
Form 
  • (1)
Product Type 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results