accessibility menu, dialog, popup

UserName

Viewing profile as user:
ClClClClClCl

CAS RN 86194-41-4

IUPAC Name: 1,2,3,4,5,6-hexachlorocyclohexane
Synonyms: lindane hexachlorocyclohexane 林旦 hexachlorocyclohexanes hexachlorzyklohexan hexachlorcyclohexan benzene hexachloride
Molecular Weight (g/mol): 290.81
Molecular Formula: C6H6Cl6
InChi Key: JLYXXMFPNIAWKQ-UHFFFAOYSA-N
SMILES: ClC1C(Cl)C(Cl)C(Cl)C(Cl)C1Cl
Grade 
  • (1)
Percent Purity 
  • (1)
Solvent or Matrix 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)