Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PESTANAL™ α-HCH-d6, ≥98% (GC), MilliporeSigma™ Supelco™

Catalog No. 111017951 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
5 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
11-101-7951 5 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 11-101-7951 Supplier MilliporeSigma Supelco Supplier No. 038115MG
Add to Cart
Add to Cart

Specifications

CAS 86194-41-4
Product Line PESTANAL
Linear Formula C6D6 Cl6
MDL Number MFCD01632293
Melting Point 159°C to 160°C
Synonym alpha-1,2,3,4,5,6-Hexachlorocyclohexane-d6; alpha-BHC-d6
Type Analytical Standard
Quantity 5 mg
Formula Weight 296.87
Percent Purity ≥98% (GC)
Grade PESTANAL™ Analytical Standard
Show More Show Less

PESTANAL is a registered trademark of Sigma-Aldrich International GmbH

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.