accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 862095-91-8

IUPAC Name: 2-(2,3,4,5,6-pentafluorophenyl)-2H,5H,6H,7H-4λ⁵-pyrrolo[2,1-c][1,2,4]triazol-4-ylium; tetrafluoroboranuide
Synonyms: 2-(2,3,4,5,6-pentafluorophenyl)-5H,6H,7H-4λ⁵-pyrrolo[2,1-c][1,2,4]triazol-4-ylium tetrafluoroborate
Molecular Weight (g/mol): 362.99
Molecular Formula: C11H7BF9N3
InChi Key: KIWCIWCCQVTPOY-UHFFFAOYSA-N
SMILES: F[B-](F)(F)F.FC1=C(F)C(F)=C(N2C=[N+]3CCCC3=N2)C(F)=C1F
Molecular Weight (g/mol) 
  • (2)
  • (1)
Quantity 
  • (1)
  • (1)
  • (1)
Percent Purity 
  • (1)
Physical Form 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)