Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

6,7-Dihydro-2-pentafluorophenyl-5H-pyrrolo[2,1-c][1,2,4]triazolium Tetrafluoroborate 98.0+%, TCI America™
SDP

Catalog No. D4624200MG Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
200 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D4624200MG 200 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. D4624200MG Supplier TCI America Supplier No. D4624200MG
Add to Cart
Add to Cart

Chemical Identifiers

CAS 862095-91-8
Molecular Formula C11H7BF9N3
Molecular Weight (g/mol) 362.994
MDL Number MFCD08459335
InChI Key KIWCIWCCQVTPOY-UHFFFAOYSA-N
PubChem CID 11152694
IUPAC Name 2-(2,3,4,5,6-pentafluorophenyl)-6,7-dihydro-5H-pyrrolo[2,1-c][1,2,4]triazol-4-ium;tetrafluoroborate
SMILES [B-](F)(F)(F)F.C1CC2=NN(C=[N+]2C1)C3=C(C(=C(C(=C3F)F)F)F)F

Specifications

Melting Point 248°C
Quantity 200 mg
UN Number 1759
Formula Weight 362.99
Percent Purity ≥98.0% (HPLC)
Physical Form Crystalline Powder
Chemical Name or Material 6,7-Dihydro-2-pentafluorophenyl-5H-pyrrolo[2,1-c][1,2,4]triazolium Tetrafluoroborate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.