accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 86259-87-2

IUPAC Name: 1-phenyl-2,5,8,11-tetraoxatridecan-13-ol
Synonyms: 1-phenyl-2,5,8,11-tetraoxatridecan-13-ol
Molecular Weight (g/mol): 284.35
Molecular Formula: C15H24O5
InChi Key: QDPIVUQXPXUNLN-UHFFFAOYSA-N
SMILES: OCCOCCOCCOCCOCC1=CC=CC=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results