Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tetraethylene Glycol Monobenzyl Ether 95.0+%, TCI America™
SDP

Catalog No. T19975G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T19975G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. T19975G Supplier TCI America Supplier No. T19975G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 86259-87-2
Molecular Formula C15H24O5
Molecular Weight (g/mol) 284.35
MDL Number MFCD06797175
InChI Key QDPIVUQXPXUNLN-UHFFFAOYSA-N
Synonym 2-[2-[2-[2-(Benzyloxy)ethoxy]ethoxy]ethoxy]ethanol
PubChem CID 11076957
IUPAC Name 1-phenyl-2,5,8,11-tetraoxatridecan-13-ol
SMILES OCCOCCOCCOCCOCC1=CC=CC=C1

Specifications

Color Yellow
Quantity 5 g
Formula Weight 284.35
Percent Purity ≥95.0% (GC)
Physical Form Liquid
Chemical Name or Material Tetraethylene Glycol Monobenzyl Ether
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.