CAS RN 863127-77-9
IUPAC Name:
N-(2-chloro-6-methylphenyl)-2-({6-[4-(2-hydroxyethyl)piperazin-1-yl]-2-methylpyrimidin-4-yl}amino)-1,3-thiazole-5-carboxamide hydrate
Synonyms:
dasatinib hydrate
Molecular Weight (g/mol):
506.02
Molecular Formula:
C22H28ClN7O3S
InChi Key:
XHXFZZNHDVTMLI-UHFFFAOYSA-N
SMILES:
O.CC1=NC(NC2=NC=C(S2)C(=O)NC2=C(C)C=CC=C2Cl)=CC(=N1)N1CCN(CCO)CC1
1
–
1
of
1
results
1
Promotions Available
Thermo Scientific Chemicals Dasatinib monohydrate
Thermo Scientific Chemicals Dasatinib monohydrate
CAS: 863127-77-9 Molecular Formula: C22H26ClN7O2S·H2O Molecular Weight (g/mol): 506.02 InChI Key: XHXFZZNHDVTMLI-UHFFFAOYSA-N PubChem CID: 11540687 ChEBI: CHEBI:70839