Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific Chemicals Dasatinib monohydrate

Catalog No. AC462320010
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC462320010 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC462320010 Supplier Thermo Scientific Chemicals Supplier No. 462320010
  • $334.00
    $263.65 / Each of 1

    Save $70.35

    21% Off The following deviations and disclosures may produce prices below stated discounts but will not trigger price reductions: Price shown reflects a temporary, promotional price reduction. No other purchase or commercial obligation required to receive the reduced price. Price may change at any time. Promotional price valid on web orders only. Your contract pricing may differ. Other exclusions apply.
View all promotions
Add to Cart

missing translation for 'validMainframeMsg'

CAS: 863127-77-9 | C22H26ClN7O2S·H2O | 506.02 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 863127-77-9
Molecular Formula C22H26ClN7O2S·H2O
Molecular Weight (g/mol) 506.02
InChI Key XHXFZZNHDVTMLI-UHFFFAOYSA-N
PubChem CID 11540687
ChEBI CHEBI:70839

Specifications

Ignition Residue 0.2% max
Quantity 1 g
Solubility Information Solubility in water: partly miscible
Formula Weight 506.02
Chemical Name or Material Dasatinib monohydrate
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement
Toxic if swallowed.
May damage fertility or the unborn child.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.