accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 86393-34-2

IUPAC Name: 2,4-dichloro-5-fluorobenzoyl chloride
Synonyms: 2,4-dichloro-5-fluorobenzoyl chloride
Molecular Weight (g/mol): 227.44
Molecular Formula: C7H2Cl3FO
InChi Key: RPZXUSJCSDQNTE-UHFFFAOYSA-N
SMILES: FC1=CC(C(Cl)=O)=C(Cl)C=C1Cl

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results