Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2,4-Dichloro-5-fluorobenzoyl Chloride 98.0+%, TCI America™
SDP

Catalog No. D43805G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D43805G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. D43805G Supplier TCI America Supplier No. D43805G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 86393-34-2
Molecular Formula C7H2Cl3FO
Molecular Weight (g/mol) 227.44
MDL Number MFCD00075341
InChI Key RPZXUSJCSDQNTE-UHFFFAOYSA-N
Synonym 2,4-dichloro-5-fluoro-benzoyl chloride, 2,4-dichloro-5-fluoro benzoyl chloride, benzoyl chloride,2,4-dichloro-5-fluoro, 2,4-dichloro-5-fluorobenzoylchloride, 2,4-bis chloranyl-5-fluoranyl-benzoyl chloride
PubChem CID 2736821
IUPAC Name 2,4-dichloro-5-fluorobenzoyl chloride
SMILES FC1=CC(C(Cl)=O)=C(Cl)C=C1Cl

Specifications

Color Yellow
Boiling Point 144°C
Quantity 5 g
UN Number 3265
Formula Weight 227.44
Percent Purity ≥98.0% (GC,T)
Physical Form Liquid
Chemical Name or Material 2,4-Dichloro-5-fluorobenzoyl Chloride
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.