accessibility menu, dialog, popup

UserName

Viewing profile as user:
H2NSHCl

CAS RN 863991-95-1

IUPAC Name: 1-[2-(thiophen-2-yl)phenyl]methanamine hydrochloride
Synonyms: 1-[2-(thiophen-2-yl)phenyl]methanamine hydrochloride
Molecular Weight (g/mol): 225.73
Molecular Formula: C11H12ClNS
InChi Key: XLJIHGHEJAGJJC-UHFFFAOYSA-N
SMILES: Cl.NCC1=CC=CC=C1C1=CC=CS1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results