CAS RN 86404-04-8
IUPAC Name:
(5R)-5-[(1S)-1,2-dihydroxyethyl]-4-ethoxy-3-hydroxy-2,5-dihydrofuran-2-one
Synonyms:
ethyl ascorbic acid 3-0-ethyl ascorbic acid 3-O-ethyl ascorbic acid
Molecular Weight (g/mol):
204.18
Molecular Formula:
C8H12O6
InChi Key:
ZGSCRDSBTNQPMS-UJURSFKZSA-N
SMILES:
CCOC1=C(O)C(=O)O[C@@H]1[C@@H](O)CO
1
–
2
of
2
results
1
3-O-Ethyl-L-ascorbic Acid 98.0+%, TCI America™
CAS: 86404-04-8 Molecular Formula: C8H12O6 Molecular Weight (g/mol): 204.178 MDL Number: MFCD09261382 InChI Key: ZGSCRDSBTNQPMS-UJURSFKZSA-N PubChem CID: 150736 IUPAC Name: (2R)-2-[(1S)-1,2-dihydroxyethyl]-3-ethoxy-4-hydroxy-2H-furan-5-one SMILES: CCOC1=C(C(=O)OC1C(CO)O)O
2
Sigma Aldrich 3-o-Ethyl-L-ascorbic acid
MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.