Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich 3-o-Ethyl-L-ascorbic acid

Catalog No. CDS009203 Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
CDS009203 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. CDS009203 Supplier Sigma Aldrich Supplier No. CDS009203100MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

3-Nitro-2-butanol undergoes oxidation catalyzed by nitroalkane oxidase enzyme isolated from extracts of Fusarium oxysporum.3-Nitro-2-butanol was used to study the mechanism of oxidation of methylethylketoxime and acetoxime in the liver microsomes from rats, mice and humans.

Specifications

CAS 86404-04-8
Molecular Formula C8H12O6
Linear Formula C8H12O6
MDL Number MFCD09261382
Quantity 25 g
Molecular Weight (g/mol) 204.18
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.