accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3NHSNCl

CAS RN 864068-99-5

IUPAC Name: {[2-(3-chlorophenyl)-1,3-thiazol-4-yl]methyl}(methyl)amine
Synonyms: {[2-(3-chlorophenyl)-1,3-thiazol-4-yl]methyl}(methyl)amine
Molecular Weight (g/mol): 238.73
Molecular Formula: C11H11ClN2S
InChi Key: YHQACXKNKUPSSA-UHFFFAOYSA-N
SMILES: CNCC1=CSC(=N1)C1=CC(Cl)=CC=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results