Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

N-{[2-(3-Chlorophenyl)-1,3-thiazol-4-yl]methyl}-N-methylamine, 90%, Thermo Scientific™

Catalog No. CC46746DA
Change view
Click to view available options
Quantity:
1 g
Packaging:
Amber Glass Bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
CC46746DA 1 g Amber Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. CC46746DA Supplier Thermo Scientific Chemicals Supplier No. CC46746DA
Add to Cart

Chemical Identifiers

CAS 864068-99-5
Molecular Formula C11H11ClN2S
Molecular Weight (g/mol) 238.733
MDL Number MFCD08690268
InChI Key YHQACXKNKUPSSA-UHFFFAOYSA-N
Synonym 4-thiazolemethanamine,2-3-chlorophenyl-n-methyl, 2-3-chlorophenyl-4-methylamino methyl-1,3-thiazole, 2-3-chlorophenyl-1,3-thiazol-4-yl methyl methyl amine, 1-2-3-chlorophenyl thiazol-4-yl-n-methylmethanamine, 2-3-chlorophenyl 1,3-thiazol-4-yl methyl methylamine, 1-2-3-chlorophenyl-1,3-thiazol-4-yl-n-methylmethanamine, 1-2-3-chlorophenyl-1,3-thiazol-4-yl-n-methylmethylamine, n-2-3-chlorophenyl-1,3-thiazol-4-yl methyl-n-methylamine
PubChem CID 18525799
IUPAC Name 1-[2-(3-chlorophenyl)-1,3-thiazol-4-yl]-N-methylmethanamine
SMILES CNCC1=CSC(=N1)C2=CC(=CC=C2)Cl

Specifications

CAS Min % 89.5
CAS Max % 100.0
Assay Percent Range 90%
Packaging Amber Glass Bottle
Quantity 1 g
Formula Weight 238.74
Percent Purity 90%
Chemical Name or Material N-{[2-(3-chlorophenyl)-1, 3-thiazol-4-yl]methyl}-N-methylamine
Hazard Statement GHS H Statement:
Causes serious eye damage.
Harmful if swallowed.
Causes severe skin burns and eye damage.
Precautionary Statement GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.