accessibility menu, dialog, popup

UserName

Viewing profile as user:
FSFFFFF

CAS RN 864230-02-4

IUPAC Name: 1-fluoro-2-(pentafluoro-λ⁶-sulfanyl)benzene
Synonyms: 1-fluoro-2-(pentafluoro-λ⁶-sulfanyl)benzene
Molecular Weight (g/mol): 222.15
Molecular Formula: C6H4F6S
InChi Key: MHUHQWYOEMJKBC-UHFFFAOYSA-N
SMILES: FC1=CC=CC=C1S(F)(F)(F)(F)F

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results