Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Fluorophenylsulfur Pentafluoride 97.0+%, TCI America™
SDP

Catalog No. F08231G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
F08231G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. F08231G Supplier TCI America Supplier No. F08231G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 864230-02-4
Molecular Formula C6H4F6S
Molecular Weight (g/mol) 222.148
MDL Number MFCD18909664
InChI Key MHUHQWYOEMJKBC-UHFFFAOYSA-N
PubChem CID 12174437
IUPAC Name pentafluoro-(2-fluorophenyl)-$l^{6}-sulfane
SMILES C1=CC=C(C(=C1)F)S(F)(F)(F)(F)F

Specifications

Color Colorless
Boiling Point 94°C
Quantity 1 g
UN Number 1993
Formula Weight 222.15
Percent Purity ≥97.0% (GC)
Physical Form Liquid
Chemical Name or Material 2-Fluorophenylsulfur Pentafluoride
TSCA No
Recommended Storage Refrigerator
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.