accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3CNH3CCH3NHOOH2OH2O

CAS RN 86426-17-7

IUPAC Name: 3-(2,2,2-trimethylhydrazin-2-ium-1-yl)propanoate dihydrate
Synonyms: meldonium
Molecular Weight (g/mol): 182.22
Molecular Formula: C6H18N2O4
InChi Key: JFWLFLLRLZSBRA-UHFFFAOYSA-N
SMILES: O.O.C[N+](C)(C)NCCC([O-])=O
Physical Form 
  • (2)
Product Type 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)