accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 864266-61-5

IUPAC Name: 1-[4-(oxan-4-yloxy)phenyl]methanamine
Synonyms: 1-[4-(oxan-4-yloxy)phenyl]methanamine
Molecular Weight (g/mol): 207.27
Molecular Formula: C12H17NO2
InChi Key: QPNQGXQLNABWSW-UHFFFAOYSA-N
SMILES: NCC1=CC=C(OC2CCOCC2)C=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results